Lanthanum(III) oxalate hydrate ≥99.9%
Supplier: Thermo Scientific Chemicals
43122-22
43122-36
CAAA43122-22EA
88.16
CAD
CAAA43122-22
CAAA43122-36
Lanthanum(III) oxalate hydrate ≥99.9%
Lanthanum(III) oxalate hydrate
CAS No.: 79079-18-8
Molecular Formula: C6La2O12.xH2O
Formula Weight: 541.87(anhy)
Physical Form: Powder
Sensitivity: Hygroscopic
Melting Point: dec.
UN No.: 3288
MDL No.: MFCD00150540
Formula:
C₆La₂O₁₂.xH₂O MW: 541.87 g/mol (anhy) |
MDL Number:
MFCD00150540 CAS Number: 79079-18-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...