Silicon dioxide 30% in ethylene glycol, colloidal dispersion

Supplier: Thermo Scientific Chemicals

Synonyms: Silicon(IV) oxideSilicium (IV) oxideSilicium dioxide

43094-A7 43094-A7 43094-A1 43094-A1
CAAA43094-A7EA 717.56 CAD
CAAA43094-A7 AA43094-A7 AA43094-A1 CAAA43094-A1
Silicon dioxide 30% in ethylene glycol, colloidal dispersion
Silicium dioxide

Viscosity @ 25° 60 cps, pH 3.0, % H2O 1.0

Formula: SiO₂
MW: 60.08 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011232
CAS Number: 7631-86-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR