Silicon dioxide 30% in ethylene glycol, colloidal dispersion
Supplier: Thermo Scientific Chemicals
Synonyms:
Silicon(IV) oxide
, Silicium (IV) oxide
, Silicium dioxide
43093-A7
43093-A1
43093-A1
43093-A7
CAAA43093-A7EA
823.86
CAD
CAAA43093-A7
CAAA43093-A1
AA43093-A1
AA43093-A7
Silicon dioxide 30% in ethylene glycol, colloidal dispersion
Silicium dioxide
Viscosity @ 25° 60 cps, pH 3.0, % H2O 1.0
Formula:
SiO₂ MW: 60.08 g/mol Density: 1.3 Storage Temperature: Ambient |
MDL Number:
MFCD00011232 CAS Number: 7631-86-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...