Poly(allylamine hydrochloride)
Supplier: Thermo Scientific Chemicals
|
Warning
|
43092-09
43092-18
CAAA43092-09EA
248.06
CAD
CAAA43092-09
CAAA43092-18
Poly(allylamine hydrochloride)
Poly(allylamine hydrochloride)
Uses: polymeric primary amine. Forms hydrogels
M.W.: 120,000 to 200,000
Soluble in water
CAS No.: 71550-12-4
Molecular Formula: [CH2CH(CH2NH2.HCl)]n
Physical Form: Powder
Appearance: Yellow
Solubility: Soluble in water
MDL No.: MFCD00084396
Applications: As a polymeric primary amine. Forms hydrogels
Formula:
[CH₂CH(CH₂NH₂·HCl)]n Storage Temperature: Ambient |
MDL Number:
MFCD00084396 CAS Number: 71550-12-4 EINECS: 415-050-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...