Ruthenium(III) nitrosyl chloride, hydrate
Supplier: Thermo Scientific Chemicals
|
Warning
|
43089-03
43089-06
CAAA43089-03EA
243.32
CAD
CAAA43089-03
CAAA43089-06
Ruthenium(III) nitrosyl chloride, hydrate
Ruthenium(III) nitrosyl chloride
CAS No.: 32535-76-5
Molecular Formula: Ru(NO)Cl3.xH2O
Formula Weight: 237.44(anhy)
Physical Form: Crystalline
Melting Point: 180° dec.
MDL No.: MFCD00049765
Formula:
Ru(NO)Cl₃ Melting Pt: 180 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00049765 CAS Number: 32535-76-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...