Calcium sulfate, Drierite™ 10-20 mesh for drying
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Calcium sulphate (anhydrous) anhydrite, Gypsum, Calcium sulfate (anhydrous) anhydrite
43058-P2
43058-P5
CAAA43058-P2EA
62.88
CAD
CAAA43058-P2
CAAA43058-P5
Calcium sulfate, Drierite™ 10-20 mesh for drying
Calcium sulphate
CAS No.: 7778-18-9
Physical Form: Granular, -10+20 Mesh
Density: 2.96
Sensitivity: Hygroscopic
Appearance: White
Odor: Odorless
Merck Reference: 14,1706
MDL No.: MFCD00010912
Formula:
CaO₄S Melting Pt: 1450 °C Density: 2.96 Storage Temperature: Ambient |
MDL Number:
MFCD00010912 CAS Number: 7778-18-9 EINECS: 231-900-3 Merck Index: 14,1706 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...