Calcium sulfate, Drierite™ 6 mm (4 mesh) for drying

Supplier: Thermo Scientific Chemicals
Warning

43056-P2 43056-P5
CAAA43056-P2EA 60.36 CAD
CAAA43056-P2 CAAA43056-P5
Calcium sulfate, Drierite™ 6 mm (4 mesh) for drying
Calcium sulphate

CAS No.: 7778-18-9 Physical Form: Granular, -4+6 Mesh Sensitivity: Hygroscopic Appearance: White Odor: Odorless Merck Reference: 14,1706 MDL No.: MFCD00010912

Formula: CaO₄S
Melting Pt: 1350 °C
Density: 2.964 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010912
CAS Number: 7778-18-9
EINECS: 231-900-3
Merck Index: 14,1706

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR