Iron(III) isopropoxide 2.5% (w/v) in isopropanol (IPA)

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Iron(III) i-propoxide

43024-AA 43024-AC
CAAA43024-AAEA 65.25 CAD
CAAA43024-AA CAAA43024-AC
Iron(III) isopropoxide 2.5% (w/v) in isopropanol (IPA)
Iron(III) isopropoxide

CAS No.: 14995-22-3 Molecular Formula: Fe[OCH(CH3)2]3 Formula Weight: 233.11 Flash Point: 12°(54°F) Physical Form: Liquid, Packaged under Argon Density: 0.79 Sensitivity: Moisture Sensitive Appearance: Red-brown Odor: Alcohol-like Boiling Point: 82° UN No.: 1219 MDL No.: MFCD00070436

Formula: C₉H₂₁FeO₃
MW: 233.109 g/mol
Boiling Pt: 82 °C
Density: 0.79
Flash Pt: 12 °C (54 °F)
Storage Temperature: Ambient
MDL Number: MFCD00070436
CAS Number: 14995-22-3
UN: 1219
ADR: 3,II

Order Now

Specification Test Results

Form Liquid, Packaged under Argon

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR