Sodium nitrite standardised solution 0.1 M in aqueous solution

Supplier: Thermo Scientific Chemicals

Synonyms: Diazotizing salts

43015-AP 43015-AP
CAAA43015-APEA 141.25 CAD
CAAA43015-AP AA43015-AP
Sodium nitrite standardised solution 0.1 M in aqueous solution
Sodium nitrite
Formula: NaNO₂
MW: 69 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011118
CAS Number: 7632-00-0
EINECS: 231-555-9
UN: 3287
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR