Silicon nitride α-phase ≥95%

Supplier: Thermo Scientific Chemicals

Synonyms: Silicon(IV) nitride

42949-18 42949-30 42949-A1
CAAA42949-18EA 82.79 CAD
CAAA42949-18 CAAA42949-30 CAAA42949-A1
Silicon nitride α-phase ≥95%
Silicon nitride

Purity: 95% CAS No.: 12033-89-5 Molecular Formula: Si3N4 85% min (alpha form) Formula Weight: 140.28 Physical Form: 10 Micron Powder, S.A. 5.0m2/g Density: 3.2 Appearance: Pale gray Odor: Odorless Melting Point: 1900° subl. Merck Reference: 14,8497 MDL No.: MFCD00011230

Formula: N₄Si₃
MW: 140.28 g/mol
Melting Pt: 1900 °C (subl.)
Density: 3.2
Storage Temperature: Ambient
MDL Number: MFCD00011230
CAS Number: 12033-89-5
EINECS: 234-796-8
Merck Index: 14,8497

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR