Stainless steel (Fe:Cr:Ni:Mo; 67.5:17:13:2.5 wt%), powder -40+80 mesh Type 316
Supplier: Thermo Scientific Chemicals
42944-30
42944-A1
CAAA42944-30EA
85.16
CAD
CAAA42944-30
CAAA42944-A1
Stainless steel (Fe:Cr:Ni:Mo; 67.5:17:13:2.5 wt%), powder -40+80 mesh Type 316
Stainless steel
Molecular Formula: Fe:Cr:Ni:Mo; 67.5:17:13:2.5 wt%
MDL No.: MFCD00148499
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...