Lithium isopropoxide ≥99.9% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
42930-04
42930-09
CAAA42930-04EA
123.56
CAD
CAAA42930-04
CAAA42930-09
Lithium isopropoxide ≥99.9% (metals basis)
Lithium isopropoxide
Purity: 99.9%
CAS No.: 2388-10-5
Molecular Formula: C3H7LiO
Formula Weight: 66.03
Physical Form: Powder, Packaged under argon
Sensitivity: Air and Moisture Sensitive
Appearance: White to pale yellow
Beilstein Reference: 3618614
UN No.: 3206
MDL No.: MFCD00048286
Formula:
C₃H₇LiO MW: 66.028 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00048286 CAS Number: 2388-10-5 UN: 3206 ADR: 4.2,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...