Gadolinium(III) sulfate octahydrate ≥99.9% (REO, rare earth oxide basis), REacton®
Supplier: Thermo Scientific Chemicals
|
Warning
|
42869-09
42869-18
CAAA42869-09EA
181.7
CAD
CAAA42869-09
CAAA42869-18
Gadolinium(III) sulfate octahydrate ≥99.9% (REO, rare earth oxide basis), REacton®
Gadolinium (III) sulphate octahydrate
Applications: Cryogenic research
Becomes anhydrous at 400°, dec. at 500°
Soluble in cold water
Purity: 99.9%
CAS No.: 13450-87-8
Molecular Formula: Gd2(SO4)3.8H2O
Formula Weight: 746.81 (602.68anhy)
Physical Form: Crystalline Aggregates
Density: 4.139
Solubility: Soluble in cold water
Merck Reference: 14,4326
MDL No.: MFCD00149734
Cryogenic research
Formula:
Gd₂O₁₂S₃·8H₂O Density: 4.139 Storage Temperature: Ambient |
MDL Number:
MFCD00149734 CAS Number: 13450-87-8 EINECS: 237-115-2 Merck Index: 14,4326 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...