Aluminum dihydrogen phosphate 50% (w/w) in aqueous solution

Supplier: Thermo Scientific Chemicals
Danger

42858-A1 42858-A7
CAAA42858-A1EA 142.99 CAD
CAAA42858-A1 CAAA42858-A7
Aluminum dihydrogen phosphate 50% (w/w) in aqueous solution
Aluminum dihydrogen phosphate

CAS No.: 13530-50-2 Molecular Formula: AlH6O12P3 Formula Weight: 317.94 Physical Form: Liquid Density: 1.5 Appearance: Colorless Odor: Odorless MDL No.: MFCD00014152

Formula: AlH₆O₁₂P₃
Density: 1.5
Storage Temperature: Ambient
MDL Number: MFCD00014152
CAS Number: 13530-50-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR