Ytterbium(III) chloride ≥35% (w/w) in aqueous solution , 99.9% (REO, rare earth oxide basis)

Supplier: Thermo Scientific Chemicals

42816-AD 42816-AK 42816-K2
CAAA42816-ADEA 82.63 CAD
CAAA42816-AD CAAA42816-AK CAAA42816-K2
Ytterbium(III) chloride ≥35% (w/w) in aqueous solution , 99.9% (REO, rare earth oxide basis)
Ytterbium (III) chloride

pH 2.5


Purity: 99.9% CAS No.: 10361-91-8 Molecular Formula: YbCl3 Formula Weight: 279.40(anhy) Physical Form: Liquid Density: 1.48 MDL No.: MFCD00011288

Formula: Cl₃Yb
Density: 1.48
Storage Temperature: Ambient
MDL Number: MFCD00011288
CAS Number: 10361-91-8
EINECS: 233-800-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR