Cobalt(II) isopropoxide
Supplier: Thermo Scientific Chemicals
|
Warning
|
42810-03
42810-06
CAAA42810-03EA
147.57
CAD
CAAA42810-03
CAAA42810-06
Cobalt(II) isopropoxide
Cobalt (II) isopropoxide
CAS No.: 60406-95-3
Molecular Formula: Co[OCH(CH3)2]2
Formula Weight: 178.11
Physical Form: Powder, Ampouled under argon
Sensitivity: Moisture Sensitive
MDL No.: MFCD00799075
Formula:
C₆H₁₄CoO₂ Storage Temperature: Ambient |
MDL Number:
MFCD00799075 CAS Number: 60406-95-3 UN: 3181 ADR: 4.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...