Tropaeolin O 0.04% (w/v) in aqueous solution
Supplier: Thermo Scientific Chemicals
42772-AE
42772-AP
CAAA42772-AEEA
50.47
CAD
CAAA42772-AE
CAAA42772-AP
Tropaeolin O 0.04% (w/v) in aqueous solution
Tropaeolin O
Transition interval: pH 11.0 (yellow) to pH 12.7 (orange)
Molecular Formula: C12H9N2NaO5S
Formula Weight: 316.27
Physical Form: Liquid
MDL No.: MFCD00007499
Formula:
C₁₂H₁₀N₂O₅S Storage Temperature: Ambient |
MDL Number:
MFCD00007499 CAS Number: 547-57-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...