Methylene blue 1% (w/v) in aqueous solution

Supplier: Thermo Scientific Chemicals

42771-AE 42771-AP
CAAA42771-AEEA 52.14 CAD
CAAA42771-AE CAAA42771-AP
Methylene blue 1% (w/v) in aqueous solution
Methylene blue

Molecular Formula: C16H18ClN3S Formula Weight: 319.86 Physical Form: Liquid MDL No.: MFCD00012111

Formula: C₁₆H₁₈ClN₃S
Storage Temperature: Ambient
MDL Number: MFCD00012111
CAS Number: 61-73-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR