Zirconium oxide polymeric precursor

Supplier: Thermo Scientific Chemicals
Danger

42766-14 42766-22
CAAA42766-14EA 181.7 CAD
CAAA42766-14 CAAA42766-22 CAAA42766-36
Zirconium oxide polymeric precursor
Zirconium oxide

Soluble in alcohols


Applications: Preparation of zirconium oxide films, solid electrolyte


Applications: Preparation of zirconium oxide films, solid electrolyte

Formula: OZr
MW: 107.22
Storage Temperature: Ambient
MDL Number: MFCD00011310
CAS Number: 12036-01-0
UN: 1993
ADR: 3 III

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR