Silica Gel, TLC High Purity Grade, With Gypsum Binder, Thermo Scientific

Supplier: Thermo Scientific Chemicals

42764-30 42764-A1
CAAA42764-18EA 51.65 CAD
CAAA42764-18 CAAA42764-30 CAAA42764-A1
Silica Gel, TLC High Purity Grade, With Gypsum Binder, Thermo Scientific
Chromatography Sorbents SPE Sorbents

This Silica gel with gypsum binder and fluorescent indicator is primarily used in plate preparation of thin layer chromatography.


  • CAS No: 63231-67-4
  • MDL No: MFCD00011232
  • Solubility: Insoluble in water

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR