Silica gel 60 0.060-0.2mm (70-230 mesh), with fluorescent indicator

Supplier: Thermo Scientific Chemicals
Warning

42757-18 42757-30 42757-A1 42757-A7
CAAA42757-18EA 39.18 CAD
CAAA42757-18 CAAA42757-30 CAAA42757-A1 CAAA42757-A7
Silica gel 60 0.060-0.2mm (70-230 mesh), with fluorescent indicator
Silica gel

pH 6.5-7.5


Applications: Dry column chromatography without applied pressure (gravity pulled)


Applications: Dry column chromatography without applied pressure (gravity pulled)

Formula: SiO₂
Storage Temperature: Ambient
MDL Number: MFCD00011232
CAS Number: 63231-67-4
EINECS: 231-545-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR