Silica gel, TLC high purity grade, without binder, Thermo Scientific
Supplier: Thermo Scientific Chemicals
42756-30
42756-A1
42756-A7
CAAA42756-18EA
88.31
CAD
CAAA42756-18
CAAA42756-30
CAAA42756-A1
CAAA42756-A7
Silica gel, TLC high purity grade, without binder, Thermo Scientific
Chromatography Sorbents
TLC Sorbents
This TLC high purity grade silica gel without binder is used for thin layer chromatography plate preparation.
- Incompatible with oxidizing agents
- Soluble in alkalies
- Insoluble in water