Samarium cobalt (33% Sm), REacton®
Supplier: Thermo Scientific Chemicals
|
Danger
|
42732-18
42732-30
CAAA42732-18EA
137.78
CAD
CAAA42732-18
CAAA42732-30
Samarium cobalt (33% Sm), REacton®
Samarium cobalt
CAS No.: 12017-68-4
Molecular Formula: SmCo5, Intermetallic, Magnet Alloy Additive
Physical Form: Fine Powder
Sensitivity: Air and Moisture Sensitive
Odor: Odorless
UN No.: 3089
MDL No.: MFCD02091682
Formula:
CoSm Storage Temperature: Ambient |
MDL Number:
MFCD02091682 CAS Number: 12017-68-4 EINECS: 234-625-7 UN: 3089 ADR: 4.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...