Calcium-3-hydroxy-3-methylbutyrate hydrate ≥97% (by titrimetric analysis), white powder
Supplier: Thermo Scientific Chemicals
|
Warning
|
42721-18
42721-30
42721-A1
CAAA42721-18EA
131.14
CAD
CAAA42721-18
CAAA42721-30
CAAA42721-A1
Calcium-3-hydroxy-3-methylbutyrate hydrate ≥97% (by titrimetric analysis), white powder
Calcium-3-hydroxy-3-methylbutyrate hydrate
Purity: 97+%
CAS No.: 135236-72-5
Molecular Formula: C10H18CaO6.xH2O
Formula Weight: 274.33(anhy)
Physical Form: Powder
Appearance: White
MDL No.: MFCD01318562
Formula:
C₁₀H₁₈CaO₆·H₂O Storage Temperature: Ambient |
MDL Number:
MFCD01321171 CAS Number: 135236-72-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...