Tetramethylammonium formate 30% (w/w) in aqueous solution
Supplier: Thermo Scientific Chemicals
42710-AE
42710-AP
CAAA42710-AEEA
130.98
CAD
CAAA42710-AE
CAAA42710-AP
Tetramethylammonium formate 30% (w/w) in aqueous solution
Tetramethylammonium formate
CAS No.: 59138-84-0
Molecular Formula: C5H13NO2
Formula Weight: 119.16
Physical Form: Liquid
Beilstein Reference: 3916177
MDL No.: MFCD00054402
Formula:
C₅H₁₃NO₂ MW: 119.164 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00054402 CAS Number: 59138-84-0 EINECS: 261-622-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...