Indium tin oxide ≥99.99% (metals basis) for vacuum deposition

Supplier: Thermo Scientific Chemicals
Warning

42677-06 42677-14 42677-22
CAAA42677-06EA 143.62 CAD
CAAA42677-06 CAAA42677-14 CAAA42677-22
Indium tin oxide ≥99.99% (metals basis) for vacuum deposition
Indium tin oxide

Purity: 99.99% CAS No.: 50926-11-9 Molecular Formula: In2O3:SnO2; 90:10 wt% Physical Form: 3-12mm Pieces MDL No.: MFCD00171662

Formula: In₂O₃·(SnO₂)x
Storage Temperature: Ambient
MDL Number: MFCD00171662
CAS Number: 50926-11-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR