Dihydroxybis(ammonium lactato)titanium(IV) 50% (w/w) in aqueous solution
Supplier: Thermo Scientific Chemicals
|
Warning
|
42676-30
42676-A1
42676-A7
CAAAA42676-30EA
81.05
CAD
CAAAA42676-30
CAAA42676-A1
CAAA42676-A7
Dihydroxybis(ammonium lactato)titanium(IV) 50% (w/w) in aqueous solution
Dihydroxybis(ammonium lactato)titanium(IV)
Applications: Water soluble titanium complex
pH 7-8
CAS No.: 65104-06-5
Molecular Formula: C6H18N2O8Ti
Formula Weight: 294.10
Flash Point: 47°(118°F)
Physical Form: Liquid
Density: 1.21
Refractive Index: 1.4380
Appearance: Pale yellow
UN No.: 1993
MDL No.: MFCD00072736
Applications: Water soluble titanium complex
Formula:
C₆H₂₂N₂O₈Ti Density: 1.21 Flash Pt: 47 °C (118 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00072736 CAS Number: 65104-06-5 EINECS: 247-428-6 UN: 1993 ADR: 3,III |
Specification Test Results
Form | Liquid |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...