Poly(styrene sulfonic acid), M.W. M.W. 75,000 30% (w/v) in aqueous solution
Supplier: Thermo Scientific Chemicals
|
Danger
|
Poly(styrene sulfonic acid) is used in water softening and to remove hard ions like calcium and magnesium. In the medical field, it is used as potassium binders in acute and chronic kidney disease. Its sodium salt is used as a superplastifier in cement, as a dye improving agent for cotton, as proton exchange membranes in fuel cell applications. It acts as a solid acid catalyst in organic synthesis. It is an antistatic resin used for electrographic and electrophotographic substrates.
CAS No.: 28210-41-5
Molecular Formula: [-CH2CH(C6H4SO3H)-]n
Physical Form: Liquid
Density: 1.1
Appearance: Amber
UN No.: 2584
MDL No.: MFCD00165973
Formula:
[-CH₂CH(C₆H₄SO₃H)-]n Density: 1.1 Storage Temperature: Ambient |
MDL Number:
MFCD00165973 CAS Number: 28210-41-5 UN: 2586 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...