Iodine monochloride ∼0.22 N in glacial acetic acid

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Iodochloride

42647-AP 42647-K2 42647-K7
CAAA42647-APEA 152.47 CAD
CAAA42647-AP CAAA42647-K2 CAAA42647-K7
Iodine monochloride ∼0.22 N in glacial acetic acid
Iodine monochloride

For determination of iodine absorption number of fats


Applications: For determination of iodine absorption number of fats


Applications: For determination of iodine absorption number of fats

Formula: ClI
MW: 162.36 g/mol
Boiling Pt: 97.3
Melting Pt: 26
Storage Temperature: Ambient
MDL Number: MFCD00011354
CAS Number: 7790-99-0
EINECS: 232-236-7
UN: 2920
ADR: 8 II
Merck Index: 13,05039

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR