Pyrite (FeS₂) naturally occurring mineral, grains ∼1.5-4.8mm (0.06-0.19in)

Supplier: Thermo Scientific Chemicals

42633-03 42633-06 42633-14
CAAA42633-03EA 50.24 CAD
CAAA42633-03 CAAA42633-06 CAAA42633-14
Pyrite (FeS₂) naturally occurring mineral, grains ∼1.5-4.8mm (0.06-0.19in)
Pyrite (FeS₂)

For grain mounts, XRD and microprobe standards, identification of unknown minerals
Hardness: 6-6.5


CAS No.: 1309-36-0 Molecular Formula: FeS2 Formula Weight: 119.98 Density: 5.02 Refractive Index: 3.08 MDL No.: MFCD00064690


Applications: For grain mounts, XRD and microprobe standards, identification of unknown minerals

Formula: FeS₂
Density: 5.02
Storage Temperature: Ambient
MDL Number: MFCD00064690
CAS Number: 1309-36-0
EINECS: 215-167-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR