Polyethylene ∼ 400 micron, low density
Supplier: Thermo Scientific Chemicals
42607-0E
42607-06
42607-14
42607-22
42607-36
CAAAA42607-0EEA
1188.16
CAD
CAAAA42607-0E
CAAA42607-06
CAAA42607-14
CAAA42607-22
CAAA42607-36
Polyethylene ∼ 400 micron, low density
Polyethylene (CAS 9002-88-4)
CAS No.: 9002-88-4
Molecular Formula: (CH2CH2)n
Physical Form: Powder
Density: 0.92
Odor: Odorless
MDL No.: MFCD00084423
Formula:
(CH₂CH₂)n Density: 0.92 Storage Temperature: Ambient |
MDL Number:
MFCD00084423 CAS Number: 9002-88-4 Merck Index: 14,7567 |
Specification Test Results
Form | Powder |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...