Dioctyldichlorosilane 97%

Supplier: Thermo Scientific Chemicals
Danger

42599-09 42599-18
CAAA42599-09EA 140.78 CAD
CAAA42599-09 CAAA42599-18
Dioctyldichlorosilane 97%
Dioctyldichlorosilane

Purity: 97% CAS No.: 18416-07-4 Molecular Formula: C16H34Cl2Si Formula Weight: 325.44 Physical Form: Liquid Density: 0.940 Appearance: Colorless to pale yellow Boiling Point: 145°/0.2mm UN No.: 2987 MDL No.: MFCD01074472

Formula: C₁₆H₃₄Cl₂Si
Boiling Pt: 145 °C (0.2 mmHg)
Density: 0.940
Storage Temperature: Ambient
MDL Number: MFCD01074472
CAS Number: 18416-07-4
UN: 2987
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR