Polyaniline, emeraldine base

Supplier: Thermo Scientific Chemicals

42581-14 42581-03 42581-06
CAAAA42581-14EA 1191.32 CAD
CAAAA42581-14 CAAA42581-03 CAAA42581-06
Polyaniline, emeraldine base
Polyaniline, emeraldine base

Applications: Conductive polymerPolyaniline is used in chemical vapor sensors, supercapacitors and biosensors. It is also useful for manufacture of electrical conducting yarns, antistatic coatings, electromagnetic shielding, flexible electrodes and printed circuit board. Further, it is used in antistatic and ESD coatings and corrosion protection.


CAS No.: 25233-30-1 Physical Form: Powder Appearance: Dark brown Melting Point: 200° dec. MDL No.: MFCD00241446


Applications: Conductive polymer

Formula: C₃H₅BrClN₃
Melting Pt: >200 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00241446
CAS Number: 25233-30-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR