Oxalate, Ion Chromatography Standard Solution, Specpure®, C_2O4-2 1000 µg/ml, Thermo Scientific
Supplier: Thermo Scientific Chemicals
42568-AP
42568-AE
CAAAA42568-APEA
353.92
CAD
CAAAA42568-AP
CAAA42568-AE
Oxalate, Ion Chromatography Standard Solution, Specpure®, C_2O4-2 1000 µg/ml, Thermo Scientific
Standards
Anionic Standards
Oxalate Standards
Guaranteed stable for six months from the ship date
Molecular Formula: Na2C2O4 in H2O
Physical Form: Liquid
Odor: Odorless
MDL No.: MFCD00012465
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...