Iodide, Ion chromatography standard solution, Specpure®, I- 1000 µg/ml, Thermo Scientific
Supplier: Thermo Scientific Chemicals
42567-AP
42567-AE
CAAAA42567-APEA
358.66
CAD
CAAAA42567-AP
CAAA42567-AE
Iodide, Ion chromatography standard solution, Specpure®, I- 1000 µg/ml, Thermo Scientific
Standards
Anionic Standards
Iodide Standards
Molecular Formula: NaI in H2O
Physical Form: Liquid
MDL No.: MFCD00011355
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...