Formate, Ion chromatography standard solution, Specpure®, HCO₂- 1000 µg/ml, Thermo Scientific
Supplier: Thermo Scientific Chemicals
42566-AP
42566-AE
CAAAA42566-APEA
363.4
CAD
CAAAA42566-AP
CAAA42566-AE
Formate, Ion chromatography standard solution, Specpure®, HCO₂- 1000 µg/ml, Thermo Scientific
Standards
Anionic Standards
Formate Standards
Guaranteed stable for six months from the ship date
Molecular Formula: HCO₂Na in H₂O
Physical Form: Liquid
MDL No.: MFCD00003297
Keep refrigerated
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...