Chromate, Ion Chromatography Standard Solution, Specpure®, CrO₄-2 1000µg/ml, Thermo Scientific
Supplier: Thermo Scientific Chemicals
42564-AP
42564-AE
CAAAA42564-APEA
368.14
CAD
CAAAA42564-AP
CAAA42564-AE
Chromate, Ion Chromatography Standard Solution, Specpure®, CrO₄-2 1000µg/ml, Thermo Scientific
Standards
Anionic Standards
Chromate(VI) Standards
Molecular Formula: K2CrO4 in H2O
Physical Form: Liquid
Appearance: Yellow
Odor: Odorless
MDL No.: MFCD00011368
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...