Acetate, Ion chromatography standard solution, Specpure®, CH₃CO₂⁻ 1000 µg/ml, Thermo Scientific
Supplier: Thermo Scientific Chemicals
42563-AP
42563-AE
CAAAA42563-APEA
357.08
CAD
CAAAA42563-AP
CAAA42563-AE
Acetate, Ion chromatography standard solution, Specpure®, CH₃CO₂⁻ 1000 µg/ml, Thermo Scientific
Standards
Anionic Standards
Acetate Standards
Guaranteed stable for six months from the ship date
Molecular Formula: CH3CO2Na in H2O
Physical Form: Liquid
Odor: Odorless
MDL No.: MFCD00282007
Keep Refrigerated
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...