Total Organic Carbon (TOC), Standard Solution, Specpure®, 1000µg/ml, Thermo Scientific
Supplier: Thermo Scientific Chemicals
42562-AE
42562-AP
CAAA42562-AEEA
232.84
CAD
CAAA42562-AE
CAAA42562-AP
Total Organic Carbon (TOC), Standard Solution, Specpure®, 1000µg/ml, Thermo Scientific
Standards
Environmental Analysis Standards
Standard for TOC based on potassium hydrogen phthalate (KHC₈H₄O₄).
Physical Form: Liquid
MDL No.: MFCD00133992
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...