Dihydrogen hexabromoosmate(IV) hydrate ≥99.9% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
42527-03
42527-06
CAAA42527-01EA
166.94
CAD
CAAA42527-01
CAAA42527-03
CAAA42527-06
Dihydrogen hexabromoosmate(IV) hydrate ≥99.9% (metals basis)
Dihydrogen hexabromoosmate(IV) hydrate
Purity: 99.9%
Molecular Formula: H2OsBr6.xH2O
Formula Weight: 352.02(anhy)
Physical Form: Crystalline
Appearance: Grey-black
Melting Point: dec.
UN No.: 3260
MDL No.: MFCD01863474
Formula:
H₂OsBr₆·xH₂O MW: 352.02 g/mol (anhy) Melting Pt: Dec. Storage Temperature: Ambient |
MDL Number:
MFCD01863474 UN: 3260 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...