Lanthanum(III) 2-methoxyethoxide 5% (w/v) in 2-methoxyethanol , 99.9% (REO, rare earth oxide basis)

Supplier: Thermo Scientific Chemicals
Danger

42522-AC 42522-AE
CAAA42522-ACEA 480.32 CAD
CAAA42522-AC CAAA42522-AE
Lanthanum(III) 2-methoxyethoxide 5% (w/v) in 2-methoxyethanol , 99.9% (REO, rare earth oxide basis)
Lanthanum(III) 2-methoxyethoxide

Preparation of lanthanum oxide thin films by the Sol-Gel method


Purity: 99.9% CAS No.: 115685-56-8 Molecular Formula: C9H21LaO6 Formula Weight: 364.17 Flash Point: 40°(104°F) Physical Form: Liquid Density: 0.96 Sensitivity: Moisture Sensitive Appearance: Yellow Boiling Point: 124° UN No.: 1188 MDL No.: MFCD00210619


Applications: Preparation of lanthanum oxide thin films by the Sol-Gel method

Formula: C₉H₂₁LaO₆
MW: 364.17 g/mol
Boiling Pt: 124 °C
Density: 0.96
Flash Pt: 40 °C (104 °F)
Storage Temperature: Ambient
MDL Number: MFCD00210619
CAS Number: 115685-56-8
UN: 1188
ADR: 3 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR