Polydimethylsiloxane, trimethylsiloxy terminated, M.W. 2000

Supplier: Thermo Scientific Chemicals

42499-AD 42499-AK 42499-K2
CAAA42499-ADEA 47.4 CAD
CAAA42499-AD CAAA42499-AK CAAA42499-K2
Polydimethylsiloxane, trimethylsiloxy terminated, M.W. 2000
Polydimethylsiloxane, trimethylsiloxy terminated

Viscosity 20 centistokes


CAS No.: 9016-00-6 Molecular Formula: (CH3)3SiO(Si(CH3)2O)nSi(CH3)3 Flash Point: 232°(450°F) Physical Form: Liquid Density: 0.966 Refractive Index: 1.4000 Appearance: Colorless Odor: Odorless Melting Point: -40° Boiling Point: 205° MDL No.: MFCD00148360

Formula: (CH₃)₃SiO(Si(CH₃)₂O)nSi(CH₃)₃
Boiling Pt: >205 °C
Melting Pt: < –40 °C
Density: 0.966
Flash Pt: 232 °C (450 °F)
Storage Temperature: Ambient
MDL Number: MFCD00148360
CAS Number: 9016-00-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR