Polydimethylsiloxane, trimethylsiloxy terminated, M.W. 1250
Supplier: Thermo Scientific Chemicals
42498-AD
42498-AK
42498-K2
CAAA42498-ADEA
45.66
CAD
CAAA42498-AD
CAAA42498-AK
CAAA42498-K2
Polydimethylsiloxane, trimethylsiloxy terminated, M.W. 1250
Polydimethylsiloxane, trimethylsiloxy terminated
Viscosity 10 centistokes
CAS No.: 9016-00-6
Molecular Formula: (CH3)3SiO(Si(CH3)2O)nSi(CH3)3
Flash Point: 263°(505°F)
Physical Form: Liquid
Density: 0.935
Refractive Index: 1.3990
Appearance: Colorless
Odor: Odorless
Melting Point: -40°
Boiling Point:
Formula:
(CH₃)₃SiO(Si(CH₃)₂O)nSi(CH₃)₃ Boiling Pt: >205 °C Melting Pt: –40 °C Density: 0.935 Flash Pt: 263 °C (505 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00148360 CAS Number: 9016-00-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...