Polydimethylsiloxane, trimethylsiloxy terminated, M.W. 237
Supplier: Thermo Scientific Chemicals
|
Warning
|
42496-AD
42496-AK
42496-K2
CAAA42496-ADEA
90.38
CAD
CAAA42496-AD
CAAA42496-AK
CAAA42496-K2
Polydimethylsiloxane, trimethylsiloxy terminated, M.W. 237
Polydimethylsiloxane, trimethylsiloxy terminated
Viscosity 1.0 centistokes
CAS No.: 9016-00-6
Molecular Formula: (CH3)3SiO(Si(CH3)2O)nSi(CH3)3
Flash Point: 39°(102°F)
Physical Form: Liquid
Density: 0.820
Refractive Index: 1.3825
Appearance: Colorless
Odor: Odorless
Melting Point: -86°
Boiling Point: 152-153°
UN No.: 1993
MDL No.: MFCD00148360
Formula:
(CH₃)₃SiO(Si(CH₃)₂O)nSi(CH₃)₃ Boiling Pt: 152…153 °C Melting Pt: –86 °C Density: 0.820 Flash Pt: 39 °C (102 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00148360 CAS Number: 9016-00-6 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...