Polydimethylsiloxane, trimethylsiloxy terminated, M.W. 17,000
Supplier: Thermo Scientific Chemicals
42492-AD
42492-AK
CAAA42492-ADEA
46.45
CAD
CAAA42492-AD
CAAA42492-AK
Polydimethylsiloxane, trimethylsiloxy terminated, M.W. 17,000
Polydimethylsiloxane, trimethylsiloxy terminated
Viscosity 500 centistokes
CAS No.: 9016-00-6
Molecular Formula: (CH3)3SiO(Si(CH3)2O)nSi(CH3)3
Flash Point: 315°(599°F)
Physical Form: Viscous liquid
Density: 0.966
Refractive Index: 1.4033
Appearance: Colorless
Odor: Odorless
Melting Point:
Formula:
(CH₃)₃SiO(Si(CH₃)₂O)nSi(CH₃)₃ Boiling Pt: 205 °C Melting Pt: < –40 °C Density: 0.966 Flash Pt: 315 °C (599 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00148360 CAS Number: 9016-00-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...