Polydimethylsiloxane, trimethylsiloxy terminated, M.W. 14,000

Supplier: Thermo Scientific Chemicals

42490-AD 42490-AK 42490-K2
CAAA42490-ADEA 46.45 CAD
CAAA42490-AD CAAA42490-AK CAAA42490-K2
Polydimethylsiloxane, trimethylsiloxy terminated, M.W. 14,000
Polydimethylsiloxane, trimethylsiloxy terminated

Viscosity 350 centistokes


CAS No.: 9016-00-6 Molecular Formula: (CH3)3SiO(Si(CH3)2O)nSi(CH3)3 Flash Point: 315°(599°F) Physical Form: Viscous liquid Density: 0.970 Refractive Index: 1.4031 Appearance: Colorless Odor: Odorless Melting Point: -40° Boiling Point: 205° MDL No.: MFCD00148360

Formula: (CH₃)₃SiO(Si(CH₃)₂O)nSi(CH₃)₃
Boiling Pt: >205 °C
Melting Pt: < –40 °C
Density: 0.970
Flash Pt: 315 °C (599 °F)
Storage Temperature: Ambient
MDL Number: MFCD00148360
CAS Number: 9016-00-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR