Inhibitor removal resin
Supplier: Thermo Scientific Chemicals
42489-14
42489-22
42489-36
CAAAA42489-14EA
52.46
CAD
CAAAA42489-14
CAAA42489-22
CAAA42489-36
Inhibitor removal resin
Inhibitor removal resin
Applications: For the removal of phenolic inhibitors: hydroquinone, hydroquinone monomethyl ether, tert-butylcatechol
CAS No.: 9003-70-7
Physical Form: Crystalline
Odor: Odorless
MDL No.: MFCD00081548
Applications: For the removal of phenolic inhibitors: hydroquinone, hydroquinone monomethyl ether, tert-butylcatechol
Storage Temperature:
Ambient |
MDL Number:
MFCD00081548 CAS Number: 9003-70-7 |
Specification Test Results
Form | Crystalline |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...