Hafnium(IV) acetylacetonate 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Hafnium(IV) 2,4-pentanedionate

42463-03 42463-06 42463-14
CAAA42463-03EA 68.89 CAD
CAAA42463-03 CAAA42463-06 CAAA42463-14
Hafnium(IV) acetylacetonate 97%
Hafnium (IV) acetylacetonate

Sublimation 82°/0.001mm


Purity: 97% CAS No.: 17475-67-1 Molecular Formula: C20H28O8Hf Formula Weight: 574.93 Physical Form: Powder Odor: Characteristic MDL No.: MFCD00044997

Formula: C₂₀H₂₈O₈Hf
MW: 574.922 g/mol
Melting Pt: 193 °C
Storage Temperature: Ambient
MDL Number: MFCD00044997
CAS Number: 17475-67-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR