3-Mercaptopropyl(dimethoxy)methylsilane ≥95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-Mercaptopropylmethyldimethoxysilane, 3-(Dimethoxymethylsilyl)-1-propanethiol

42426-AD 42426-AK
CAAA42426-ABEA 37.08 CAD
CAAA42426-AB CAAA42426-AD CAAA42426-AK CAAA42426-K2
3-Mercaptopropyl(dimethoxy)methylsilane ≥95%
3-Mercaptopropyl(dimethoxy)methylsilane

Purity: 95% CAS No.: 31001-77-1 Molecular Formula: C6H16O2SSi Formula Weight: 180.34 Flash Point: 93°(199°F) Physical Form: Liquid Density: 1.000 Refractive Index: 1.4500 Sensitivity: Moisture Sensitive Appearance: Colorless Boiling Point: 96°/30mm MDL No.: MFCD00053923

Formula: C₆H₁₆O₂SSi
MW: 180.34 g/mol
Boiling Pt: 96 °C (30 mmHg)
Density: 1,000
Flash Pt: 93 °C (199 °F)
Storage Temperature: Ambient
MDL Number: MFCD00053923
CAS Number: 31001-77-1
EINECS: 250-426-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR