Chlorodimethyloctadecylsilane ≥95%
Supplier: Thermo Scientific Chemicals
|
Danger
|
42414-06
42414-14
42414-22
42414-36
CAAA42414-06EA
48.82
CAD
CAAA42414-06
CAAA42414-14
CAAA42414-22
CAAA42414-36
Chlorodimethyloctadecylsilane ≥95%
Chlorodimethyloctadecylsilane
Preparation of hydrophobic surfaces, chain terminator
Purity: 95%
CAS No.: 18643-08-8
Molecular Formula: C20H43ClSi
Formula Weight: 347.10
Flash Point: 200°(392°F)
Physical Form: Solid
Density: 0.850
Refractive Index: 1.4530
Sensitivity: Moisture Sensitive
Appearance: White
Odor: Acrid
Melting Point: 28-32°
Boiling Point: 184-186°/0.2mm
Beilstein Reference: 2246012
UN No.: 3261
MDL No.: MFCD00000504
Preparation of hydrophobic surfaces, chain terminator
Formula:
C₂₀H₄₃ClSi Boiling Pt: 184…186 °C (0.2 mmHg) Melting Pt: 28…32 °C Density: 0,850 Flash Pt: 200 °C (392 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000504 CAS Number: 18643-08-8 EINECS: 242-472-2 UN: 3261 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...